Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZIC2 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA5142713
This item is not returnable.
View return policy
Description
Additional Information: This antibody is expected to NOT cross-react with other ZIC proteins. This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:1,000.
ZIC2 (zinc finger protein of the cerebellum 2) is a transcriptional repressor and may regulate tissue specific expression of dopamine receptor D1. In mouse, knockout of Zic1 leads to aplasia of cerebellum and skeletal abnormalities. Mutation in Zic2 and Zic3 in mouse and/or human cause holoprosencephaly and heterotaxis, respectively. Holoprosencephaly is the most common structural anomaly of the human brain.
Specifications
| ZIC2 | |
| Polyclonal | |
| Unconjugated | |
| ZIC2 | |
| cerebellum zinc finger protein 2; GENA 29; HPE5; Ku; kumba; LOW QUALITY PROTEIN: zinc finger protein ZIC 2; odd-paired homolog; Zic family member 2; Zic family member 2 (odd-paired homolog, Drosophila); ZIC2; Zinc finger protein of the cerebellum 2; zinc finger protein ZIC 2; zinc finger protein Zic2 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 7546 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| O95409 | |
| ZIC2 | |
| Peptide with sequence C-HSGLSSNFNEWY. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction