Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zic2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981514
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1514 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1514 Supplier Bioss Supplier No. BS11610R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZIC2 (zinc finger protein of the cerebellum 2) is a transcriptional repressor and may regulate tissue specific expression of dopamine receptor D1. In mouse, knockout of Zic1 leads to aplasia of cerebellum and skeletal abnormalities. Mutation in Zic2 and Zic3 in mouse and/or human cause holoprosencephaly and heterotaxis, respectively. Holoprosencephaly is the most common structural anomaly of the human brain.

Specifications

Antigen Zic2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ZIC2
Gene Accession No. O95409
Gene Alias BOS_12680; cerebellum zinc finger protein 2; GENA 29; HPE5; Ku; kumba; odd-paired homolog; Zic family member 2; Zic family member 2 (odd-paired homolog, Drosophila); ZIC2; Zinc finger protein of the cerebellum 2; Zinc finger protein ZIC 2
Gene Symbols ZIC2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Zic2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7546
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.