Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZIC3 Antibody (OTI1G7), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274925G
Description
ZIC3 Monoclonal specifically detects ZIC3 in Human, Mouse samples. It is validated for Western Blot.Specifications
ZIC3 | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ZIC3 | |
Recombinant protein fragment corresponding to amino acids 242-467 of human ZIC3 (NP_003404) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse | |
IgG1 |
Western Blot | |
OTI1G7 | |
Western Blot | |
heterotaxy 1, HTXHTX1, Zic family member 3 (odd-paired homolog, Drosophila), Zinc finger protein 203, zinc finger protein ZIC 3, ZNF203Zinc finger protein of the cerebellum 3 | |
Mouse | |
Affinity Purified | |
Core ESC Like Genes, Stem Cell Markers | |
7547.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction