Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZIC3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597216
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597216 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597216 Supplier Invitrogen™ Supplier No. PA597216
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MTMLLDGGPQ FPGLGVGSFG APRHHEMPNR EPAGMGLNPF GDSTHAAAAA AAAAAFKLSP AAAHDLSSGQ SSAFTPQGSG YANALGHHHH HHHHHHHTSQ VPSYGGAASA AFNSTREFLF RQRSSGLSEA ASGGGQHGLF AGSASSLHAP AGIPEPPSYL LFPGLHEQGA GHPSPTGHVD NNQVHLGLRG ELFGRADPYR PVASPRTDPY AAGAQFPNYS PMNMNMGVNV AAHHGPGAFF; Positive Samples: MCF-7, Mouse brain, Rat brain; Cellular Location: Cytoplasm, Nucleus.

This gene encodes a member of the ZIC family of C2H2-type zinc finger proteins. This nuclear protein probably functions as a transcription factor in early stages of left-right body axis formation. Mutations in this gene cause X-linked visceral heterotaxy, which includes congenital heart disease and left-right axis defects in organs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZIC3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.38 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Zic3
Gene Accession No. O60481, Q62521
Gene Alias Bn; heterotaxy 1; HTX; HTX1; Ka; RGD1561261; VACTERLX; ZIC; Zic family member 3; Zic family member 3 (odd-paired homolog, Drosophila); ZIC1; ZIC2; Zic3; Zinc finger protein 203; zinc finger protein of the cerebellum 3; Zinc finger protein ZIC 3; ZNF201; ZNF203
Gene Symbols Zic3
Host Species Rabbit
Immunogen Recombinant Protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22773, 367944, 7547
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.