Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZIC3 Polyclonal Antibody
Rabbit Polyclonal Antibody
$589.50
Specifications
Antigen | ZIC3 |
---|---|
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5111364
|
Invitrogen™
PA5111364 |
100 μL |
Each of 1 for $589.50
|
|
|||||
Description
Immunogen sequence: HEQGAGHPSP TGHVDNNQVH LGLRGELFGR ADPYRPVA.
This gene encodes a member of the ZIC family of C2H2-type zinc finger proteins. This nuclear protein probably functions as a transcription factor in early stages of left-right body axis formation. Mutations in this gene cause X-linked visceral heterotaxy, which includes congenital heart disease and left-right axis defects in organs.Specifications
ZIC3 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O60481 | |
7547 | |
Recombinant Protein corresonding to Human ZIC3. Recombinant protein control fragment (Product #RP-100975). | |
Antibody |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Polyclonal | |
Liquid | |
IgG | |
Human | |
Zic3 | |
Bn; heterotaxy 1; HTX; HTX1; Ka; RGD1561261; VACTERLX; ZIC; Zic family member 3; Zic family member 3 (odd-paired homolog, Drosophila); ZIC1; ZIC2; Zic3; Zinc finger protein 203; zinc finger protein of the cerebellum 3; Zinc finger protein ZIC 3; ZNF201; ZNF203 | |
Zic3 | |
Primary | |
Antigen affinity chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title