Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZIC4 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA556392
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556392 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556392 Supplier Thermo Scientific Supplier No. PA556392

Rabbit Polyclonal Antibody

Immunogen sequence: LVMRKRLRLY RNTLKESSSS SGHHGPQLTA ASSPSVFPGL HEEPPQASPS RPLNGLLRLG LPGDMYARPE PFPPGPAARS DALAAAAALH GYGGMNLTVN L Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 84%.

This gene encodes a member of the ZIC family of C2H2-type zinc finger proteins. Members of this family are important during development, and have been associated with X-linked visceral heterotaxy and holoprosencephaly type 5. This gene is closely linked to the gene encoding zinc finger protein of the cerebellum 1, a related family member on chromosome 3. Heterozygous deletion of these linked genes is involved in Dandy-Walker malformation, which is a congenital cerebellar malformation. Multiple transcript variants have been identified for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZIC4
Applications Immunocytochemistry, Immunohistochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZIC4
Gene Accession No. Q8N9L1
Gene Alias Zic family member 4; ZIC4; zinc family member 4 protein HZIC4; zinc finger protein of the cerebellum 4; Zinc finger protein ZIC 4
Gene Symbols ZIC4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZIC4
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84107
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.