Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZIC5 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981982
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1982 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1982 Supplier Bioss Supplier No. BS12147R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a member of the ZIC family of C2H2-type zinc finger proteins. Members of this family are important during development. Aberrant expression of this gene is seen in medulloblastoma, a childhood brain tumor. This gene is closely linked to the gene encoding zinc finger protein of the cerebellum 4, a related family member on chromosome 3. This gene encodes a transcription factor that can bind and transactivate the apolipoprotein E gene. [provided by RefSeq, Jul 2008].

Specifications

Antigen ZIC5
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ZIC2
Gene Accession No. O60481, O95409, Q15915, Q8N9L1, Q96T25
Gene Alias 1700049L20Rik; Bn; cb807; cerebellum zinc finger protein 2; CRS6; fj36d02; GENA 29; heterotaxy 1; HPE5; HTX; HTX1; id:ibd2176; Ka; Ku; kumba; LOC485704; LOW QUALITY PROTEIN: zinc finger protein ZIC 3; Odd paired-related protein; odd-paired homolog; odd-paired related; odd-paired-like; Opa-related protein; opl; Opr; RGD1561261; VACTERLX; wu:fj36d02; wu:fj51c06; wu:fj52a10; wu:fq33c12; zgc:136736; zgc:154123; Zic; Zic family member 1; Zic family member 1 (odd-paired homolog, Drosophila); Zic family member 2; Zic family member 2 (odd-paired homolog, Drosophila); Zic family member 3; Zic family member 3 (odd-paired homolog, Drosophila); Zic family member 3 heterotaxy 1 (odd-paired homolog, Drosophila); Zic family member 4; Zic family member 4 (odd-paired homolog, Drosophila); Zic family member 5; Zic family member 5 (odd-paired homolog, Drosophila); zic protein member 1; ZIC1; ZIC2; Zic3; Zic4; ZIC5; zinc family member 4 protein HZIC4; zinc family member 5 protein; zinc finger of the cerebellum 4; zinc finger protein; Zinc finger protein 201; Zinc finger protein 203; Zinc finger protein of the cerebellum 1; Zinc finger protein of the cerebellum 2; Zinc finger protein of the cerebellum 3; Zinc finger protein of the cerebellum 4; Zinc finger protein of the cerebellum 5; Zinc finger protein ZIC 1; Zinc finger protein ZIC 2; Zinc finger protein ZIC 3; Zinc finger protein ZIC 4; Zinc finger protein ZIC 5; zinc finger protein zic1; zinc finger protein Zic5; ZNF201; ZNF203
Gene Symbols ZIC1, ZIC2, ZIC3, ZIC4, Zic5
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ZIC5.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7545, 7546, 7547, 84107, 85416
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.