Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZIK1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501984300
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4300 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4300 Supplier Bioss Supplier No. BS19266R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

May be a transcriptional repressor.

Specifications

Antigen ZIK1
Applications Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ZIK1
Gene Accession No. Q3SY52
Gene Alias ZIK1; Zinc finger protein 762; zinc finger protein interacting with K protein 1; zinc finger protein interacting with K protein 1 homolog; zinc finger protein interacting with K protein 1 homolog (mouse); Zinc finger protein interacting with ribonucleoprotein K; ZNF762
Gene Symbols ZIK1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ZIK1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284307
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.