Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Zika Virus Capsid (strain H/PF/2013) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143438
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143438 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5143438 Supplier Invitrogen™ Supplier No. PA5143438
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store as concentrated solution. Centrifuge briefly prior to opening vial. Positive control: Zika Virus-PRVABC59 infected C6/36 cells, Zika-infected Vero extracts, Zika virus Capsid-transfected 293T.

Zika virus (ZIKV) capsid proteins efficiently recruit and surround the viral RNA at the endoplasmic reticulum (ER) membrane to yield nascent viral particles The capsid proteins of Zika virus (ZIKV) interact with viral RNA, forming a nucleocapsid (NC), in the process of viral genome packaging This process is called encapsidation, and up to date, a clear notion concerning the molecular mechanisms behind this phenomenon is not fully understood [PMID: 34375636].
TRUSTED_SUSTAINABILITY

Specifications

Antigen Zika Virus Capsid (strain H/PF/2013)
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.33 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.025% Proclin 300; pH 7
Gene Alias Zika virus Capsid; ZIKV Capsid
Host Species Rabbit
Immunogen Full length Zika virus Capsid recombinant protein. (Zika virus (strain H/PF/2013)).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.