Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Zika Virus Envelope Protein Monoclonal Antibody (6932)
GREENER_CHOICE

Catalog No. PIMA533303
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA533303 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA533303 Supplier Invitrogen™ Supplier No. MA533303
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Does not cross react with the following viruses: Dengue 1-4, and Chikungunya virus.

As a member of the Flaviviridae family, ZIKV has a single positive sense RNA genome, which is initially translated as a single polyprotein which is then cleaved post-transitionally into three structural proteins (C, PrM or M, and E) and seven nonstructural (NS) proteins (NS1, NS2A, NS2B, NS3, NS4A, NS4B and NS5)(1). The glycosated NS1 forms a homodimer inside the cells and is necessary for viral replication and late in infection (2). NS1 is also secreted into the extracellular space as a hexameric lipoprotein particle, which assists the virus in immune evasion and pathogenesis by interacting with components from both innate and adaptive immune systems as well as other host factors (1). Reference 1) Song, H.; Qi, J.; Haywood, J.; Shi, Y.; Gao, G.F. (2016). Nat. Struct. Mol. Biol. 23, 456-458. 2) Suthar, M.S., Diamond, M.S. and Gale, M. Jr. (2013). Nat. Rev. Microbiol. 11, 115-128.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Zika Virus Envelope Protein
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 6932
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene Alias Zika Envelope
Host Species Mouse
Purification Method Protein A
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.