Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Zika Virus IgG Antibody, DyLight 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP261131V
This item is not returnable.
View return policy
Description
Zika Virus IgG Polyclonal antibody specifically detects Zika Virus IgG in Virus samples. It is validated for ELISASpecifications
| Zika Virus IgG | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Zika Capsid antibody was raised against a peptide corresponding to 14 amino acids near the amino terminus of human Zika Capsid. Amino Acid Squence: RIINARKEKKRRGADTS | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication | |
| Virus | |
| Purified |
| ELISA | |
| DyLight 405 | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction