Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Zika Virus IgG Antibody, DyLight 650, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP261131C
This item is not returnable.
View return policy
Description
Zika Virus IgG Polyclonal antibody specifically detects Zika Virus IgG in Virus samples. It is validated for ELISASpecifications
Zika Virus IgG | |
Polyclonal | |
50mM Sodium Borate | |
Zika Capsid antibody was raised against a peptide corresponding to 14 amino acids near the amino terminus of human Zika Capsid. Amino Acid Squence: RIINARKEKKRRGADTS | |
0.1 mL | |
Cancer, Cell Cycle and Replication | |
Virus | |
Purified |
ELISA | |
DyLight 650 | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction