Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals Zika Virus Native Protein
SDP

Catalog No. NB159139 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
0.25 mg
0.5 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB159139 0.1 mg
NB159140 0.25 mg
NB159141 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. NB159139 Supplier Novus Biologicals Supplier No. NBP3148320.1MG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Specifications

For Use With (Application) ELISA
Formulation PBS pH7.4
Quantity 0.1 mg
Storage Requirements Store at 4C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles.
Protein Zika Virus
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.