Learn More
Invitrogen™ Zika Virus NS1 Monoclonal Antibody (EA23)

Description
MA5-24582 does not cross react with the following viruses: Dengue 1-4, TBE virus, West Nile virus, Yellow Fever virus, Japanese Encephalitis virus and Chikungunya virus.
Specifications
Specifications
| Antigen | Zika Virus NS1 |
| Applications | ELISA, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | EA23 |
| Concentration | 100 μg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide; pH 7.2 |
| Gene Alias | Zika NS1; ZIKV NS1; ZIKV-NS1 |
| Host Species | Mouse |
| Immunogen | recombinant Zika Virus NS1 protien. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.