Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zika virus NS1 Mouse anti-Virus, Clone: M86640, Abnova™

Catalog No. 89033718 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-033-718 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-033-718 Supplier Abnova Corporation Supplier No. MAB14218
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against partial recombinant Zika virus NS1.

  • High specificity
  • High reactivity

Specifications

Antigen Zika virus NS1
Applications ELISA, Lateral Flow
Classification Monoclonal
Clone M86640
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant Zika virus NS1.
Dilution ELISA Lateral-Flow Immunochromatographic Assay The optimal working dilution should be determined by the end user.
Formulation In 0.015 M potassium phosphate, 0.85% NaCl, pH 7.2 (0.05% sodium azide).
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 792-1144 of Zika virus NS1.
Purification Method DEAE column chromatography
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity No cross reactivity with recombinant Dengue and Chikungunya antigens.
Target Species Virus
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1 κ
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.