Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zika Virus NS1 protein Mouse anti-Virus, Unconjugated, Clone: 110, MilliporeSigma™

Catalog No. MABF21871MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF21871MI 100 μL
MABF21872MI 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF21871MI Supplier MilliporeSigma Supplier No. MABF2187100UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Zika Virus NS1 protein Monoclonal antibody specifically detects Zika Virus NS1 protein in Virus samples. It is validated for ELISA, Flow Cytometry, Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Zika Virus NS1 protein
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 110
Conjugate Unconjugated
Gene Alias ZIka Virus NS1 protein
Host Species Mouse
Immunogen Recombinant Zika virus NS1 protein expressed in mammalian cells.
Molecular Weight of Antigen ∼47 kDa observed. Uncharacterized bands may be observed in some lysate(s).
Purification Method Protein G Purified
Quantity 100 μL
Research Discipline Inflammation and Immunology
Primary or Secondary Primary
Test Specificity Clone 110 is a mouse monoclonal antibody that specifically detects Zika virus NS1 protein.
Target Species Virus
Content And Storage Stable for 1 year at 2 to 8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.