Learn More
Zika Virus NS1 protein Mouse anti-Virus, Unconjugated, Clone: 110, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Zika Virus NS1 protein |
| Applications | ELISA, Flow Cytometry, Western Blot |
| Classification | Monoclonal |
| Clone | 110 |
| Conjugate | Unconjugated |
| Gene Alias | ZIka Virus NS1 protein |
| Host Species | Mouse |
| Immunogen | Recombinant Zika virus NS1 protein expressed in mammalian cells. |
| Molecular Weight of Antigen | ∼47 kDa observed. Uncharacterized bands may be observed in some lysate(s). |
| Purification Method | Protein G Purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.