Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Zika Virus NS2B Recombinant Rabbit Monoclonal Antibody (HL2151)
GREENER_CHOICE

Catalog No. FEMA550404
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550404 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550404 Supplier Invitrogen™ Supplier No. MA550404
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Zika Virus NS2B Recombinant Monoclonal Antibody for Western Blot, ICC/IF

Zika virus (ZIKV) outbreaks are linked to severe neurological disorders ZIKV relies on its NS2B/NS3 protease for polyprotein processing; hence, this enzyme is an attractive drug target NS2B/NS3 viral protease, which, together with host cell proteases, cleaves the viral polyprotein into the individual proteins required for viral replication [DOI: 10.1126/science aag2419].
TRUSTED_SUSTAINABILITY

Specifications

Antigen Zika Virus NS2B
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone HL2151
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Alias Zika virus Non-structural protein 2B; Zika virus NS2B; ZIKV Non-structural protein 2B; ZIKV NS2B
Host Species Rabbit
Immunogen Synthetic peptide of Zika virus NS2B protein.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.