Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Zika Virus NS4A (strain H/PF/2013) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143444
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143444 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5143444 Supplier Invitrogen™ Supplier No. PA5143444
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store as concentrated solution. Centrifuge briefly prior to opening vial.

In flaviviruses, the NS4A is an integral transmembrane protein that contributes to form virus-induced membrane curvature However, structural features of NS4A are not documented in Zika virus, and it is one of the least characterized proteins The conformational change in NS4A (residues 1-48) secondary structure upon interaction with lipid vesicles can be correlated with the disorder-function paradigm concept This change in NS4A (residues 1-48) secondary structure may suggest its implication in membrane rearrangement and replication complex formation [PMID: 32871423].
TRUSTED_SUSTAINABILITY

Specifications

Antigen Zika Virus NS4A (strain H/PF/2013)
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.35 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% Proclin 300; pH 7
Gene Alias Zika virus Non-structural protein 4A; Zika virus NS4A; ZIKV Non-structural protein 4A; ZIKV NS4A
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of Zika virus NS4A protein (Zika virus (strain H/PF/2013)). The exact sequence is proprietary.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.