Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Zika Virus NS4B (strain H/PF/2013) Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5143437
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial.
Zika virus (ZIKV) NS4B protein is a membranotropic multifunctional protein Despite its versatile functioning, its topology and dynamics are not entirely understood There is no X-ray or cryo-EM structure available for any flaviviral NS4B full-length protein [PMID: 36037701].
Specifications
Zika Virus NS4B (strain H/PF/2013) | |
Polyclonal | |
Unconjugated | |
Zika virus Non-structural protein 4B; Zika virus NS4B; ZIKV Non-structural protein 4B; ZIKV NS4B | |
Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of Zika virus NS4B protein (Zika virus (strain H/PF/2013)). The exact sequence is proprietary. | |
100 μL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Flow Cytometry, Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 20% glycerol and 0.025% Proclin 300; pH 7 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
Virus | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction