Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Zika Virus prM (strain H/PF/2013) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143443
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143443 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5143443 Supplier Invitrogen™ Supplier No. PA5143443
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store as concentrated solution. Centrifuge briefly prior to opening vial. Positive control: Zika virus prM-infected C6/36.

Zika virus has biological similarities to other flaviviruses, such as West Nile virus, Japanese encephalitis virus (JEV) and dengue virus ZIKV has an approximately 11 kb positive-stranded RNA genome, with untranslated regions flanking the 5' and 3' ends of its open reading frame The genome is translated into structural proteins [capsid (C), precursor membrane (prM), and envelope (E)] and non-structural (NS) proteins (NS1, NS2A, NS2B, NS3, NS4A, NS4B, and NS5) Precursor membrane (prM), one of the ZIKV structural proteins, plays an important role in the assembly of mature virions through cleavage of prM into M protein Recent studies have suggested that prM protein might be implicated in the neurovirulence of ZIKV Most vaccines targeting ZIKV include prM as the immunogen [PMID: 30116235].
TRUSTED_SUSTAINABILITY

Specifications

Antigen Zika Virus prM (strain H/PF/2013)
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.53 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% Proclin 300; pH 7
Gene Alias Zika virus Premembrane; Zika virus PrM; ZIKV Premembrane; ZIKV PrM
Host Species Rabbit
Immunogen Full length Zika virus prM recombinant protein. (Zika virus (strain H/PF/2013)).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.