Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Zika Virus (SPH2015) Envelope Antibody (029)

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290342
Description
Zika Virus (SPH2015) Envelope Monoclonal specifically detects Zika Virus (SPH2015) Envelope in Virus samples. It is validated for Western Blot.Specifications
| Zika Virus (SPH2015) Envelope | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Zika Virus (SPH2015) Envelope (ALU33341.1; Val593-Lys699). | |
| 50 μL | |
| Primary | |
| Virus | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 29 | |
| Western Blot 1:1000-1:5000 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Zika virus (ZIKV) (strain Zika SPH2015) ZIKV-E/Envelope protein (Domain III) | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction