Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Zika Virus (SPH2015) NS1 Antibody (03), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP305857APC
Description
Zika Virus (SPH2015) NS1 Monoclonal antibody specifically detects Zika Virus (SPH2015) NS1 in Virus samples. It is validated for ELISASpecifications
Zika Virus (SPH2015) NS1 | |
Monoclonal | |
APC | |
PBS | |
Mouse | |
Protein A purified | |
RUO | |
Virus | |
Purified |
ELISA | |
03 | |
Sandwich ELISA Capture | |
Zika NS1, ZIKV NS1, ZIKV-NS1 | |
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Zika Virus (Accession#: ALU33341.1; Val796-Leu1157). | |
0.1 mL | |
Primary | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction