Learn More
Zika Virus (SPH2015) NS1 Mouse anti-Virus, HRP, Antibody Pair, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Zika Virus (SPH2015) NS1 |
| Applications | ELISA |
| Classification | Monoclonal |
| Conjugate | HRP |
| Gene Alias | Zika NS1, ZIKV NS1, ZIKV-NS1 |
| Host Species | Mouse |
| Quantity | 15 Plates |
| Regulatory Status | RUO |
| Target Species | Virus |
| Content And Storage | Storage of components varies. See protocol for specific instructions. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.