Learn More
Zika (ZIKV) E Protein Monoclonal Antibody (ZV67), Functional Grade, In Vivo GOLD™, Leinco Technologies™

Mouse Monoclonal Antibody
Supplier: Leinco Technologies Inc Z200100MG


Description
Specificity: Clone ZV-67 binds to the Zika virus envelope (E) protein at domain III (DIII, LR). Endotoxin Level: <1.0 EU/μg as determined by the LAL method. Purity: ≥95% monomer by analytical SEC and >95% by SDS Page. Preparation: Functional grade preclinical antibodies are manufactured in an animal free facility using in vitro cell culture techniques and are purified by a multi-step process including the use of protein A or G to assure extremely low levels of endotoxins, leachable protein A or aggregates. Storage: Functional grade preclinical antibodies may be stored sterile as received at 2-8C for up to one month. For longer term storage, aseptically aliquot in working volumes without diluting and store at -80C. Avoid Repeated Freeze Thaw Cycles.
Zika (ZIKV) E Protein Monoclonal antibody specifically detects Zika (ZIKV) E Protein in Virus samples. It is validated for ELISA, Functional Assay, Neutralization, Western BlotSpecifications
Zika (ZIKV) E Protein | |
Monoclonal | |
≥5.0 mg/mL | |
PBS with no preservative; pH 7.2-7.4 | |
Mouse | |
Protein A/G | |
RUO | |
Virus | |
Antibody | |
IgG2c |
ELISA, Functional Assay, Neutralization, Western Blot | |
ZV67 | |
Functional Grade | |
Zika Envelope | |
Injection of a Mouse with ZIKV MR-766, ZIKV H/PF/2013, and ZIKV DIII | |
100 mg | |
Primary | |
4°C short term, -80°C long term | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.