Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZIM2 Antibody (OTI6D12), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274928AF350
Description
ZIM2 Monoclonal specifically detects ZIM2 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
ZIM2 | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
ZIM2 | |
Recombinant protein fragment corresponding to amino acids 1-150 and 428-527 of human ZIM2 (NP_056178) produced in E.coli. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI6D12 | |
Western Blot, Immunohistochemistry | |
zinc finger imprinted 2, zinc finger, imprinted 2, ZNF656Zinc finger protein 656 | |
Mouse | |
Affinity Purified | |
modENCODE Antibodies | |
23619.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction