Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZIM2 Antibody (OTI6D12), DyLight 488, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP274928G

Catalog No. NBP274928G


Only null left
Add to Cart

Description

Description

ZIM2 Monoclonal specifically detects ZIM2 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

ZIM2
Monoclonal
DyLight 488
50mM Sodium Borate with 0.05% Sodium Azide
ZIM2
Recombinant protein fragment corresponding to amino acids 1-150 and 428-527 of human ZIM2 (NP_056178) produced in E.coli.
0.1 mL
Primary
Human
IgG1
Western Blot, Immunohistochemistry
OTI6D12
Western Blot, Immunohistochemistry
zinc finger imprinted 2, zinc finger, imprinted 2, ZNF656Zinc finger protein 656
Mouse
Affinity Purified
modENCODE Antibodies
23619.0
Store at 4°C in the dark.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.