Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZIM2 Monoclonal Antibody (OTI5C7), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA501904

Description
In human, ZIM2 and PEG3 (GeneID:5178) are two distinct genes that share a set of 5′ exons and have a common promoter, and both genes are paternally expressed. Alternative splicing events connect the shared exons either with the remaining 4 exons unique to ZIM2, or with the remaining 2 exons unique to PEG3. This is in contrast to mouse and cow, where ZIM2 and PEG3 genes do not share exons in common, and the imprinting status of ZIM2 is also not conserved amongst mammals. Alternatively spliced transcripts encoding the same protein have been found for the human ZIM2 gene.Specifications
ZIM2 | |
Monoclonal | |
0.6 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q9NZV7 | |
ZIM2 | |
Human recombit protein fragment corresponding to amino acids 1-150 and 428-527 of human ZIM2 produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Western Blot | |
OTI5C7 | |
Unconjugated | |
ZIM2 | |
ZNF656 | |
Mouse | |
Affinity Chromatography | |
RUO | |
23619 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction