Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZIM2 Monoclonal Antibody (OTI6D12)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA525304
Description
ZIM2 Monoclonal Antibody for Western Blot, IHC (P)
In human, ZIM2 and PEG3 (GeneID:5178) are two distinct genes that share a set of 5' exons and have a common promoter, and both genes are paternally expressed. Alternative splicing events connect the shared exons either with the remaining 4 exons unique to ZIM2, or with the remaining 2 exons unique to PEG3. This is in contrast to mouse and cow, where ZIM2 and PEG3 genes do not share exons in common, and the imprinting status of ZIM2 is also not conserved amongst mammals. Alternatively spliced transcripts encoding the same protein have been found for the human ZIM2 gene.
Specifications
ZIM2 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9NZV7 | |
ZIM2 | |
Human recombinant protein fragment corresponding to amino acids 1-150 and 428-527 of ZIM2 produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
OTI6D12 | |
Unconjugated | |
ZIM2 | |
ZIM2; Zinc finger imprinted 2; zinc finger protein 656; zinc finger, imprinted 2; ZNF656 | |
Mouse | |
Affinity Chromatography | |
RUO | |
23619 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction