Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZIM3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | ZIM3 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
ZIM3 Polyclonal specifically detects ZIM3 in Human samples. It is validated for Western Blot.Specifications
ZIM3 | |
Polyclonal | |
Rabbit | |
Human | |
MGC138876, MGC138877, zinc finger imprinted 3, zinc finger protein 657, zinc finger, imprinted 3, ZNF657 | |
ZIM3 | |
IgG | |
54 kDa |
Western Blot | |
Unconjugated | |
RUO | |
NP_443114 | |
114026 | |
Synthetic peptide directed towards the middle region of human ZIM3. Peptide sequence CGKAFFQKSNLHSHQKTHSGERTYRCSECGKTFIRKLNLSLHKKTHTGQK. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title