Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZIM3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541198
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541198 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541198 Supplier Invitrogen™ Supplier No. PA541198
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: CGKAFFQKSN LHSHQKTHSG ERTYRCSECG KTFIRKLNLS LHKKTHTGQK Sequence homology: Human: 100%.

ZIM3 belongs to the krueppel C2H2-type zinc-finger protein family. It contains 11 C2H2-type zinc fingers and 1 KRAB domain. ZIM3 may be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZIM3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ZIM3
Gene Accession No. Q96PE6
Gene Alias ZIM3; Zinc finger imprinted 3; zinc finger protein 657; zinc finger, imprinted 3; ZNF657
Gene Symbols ZIM3
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human ZIM3.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114026
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.