Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc 2,4-pentanedionate monohydrate

Catalog No. p-7049022
Click to view available options
Quantity:
50 g
250 g

CAS: 14363-15-6 | C10H16O5Zn | 281.61 g/mol

Zinc Metal acetylacetonates are used as catalysts for polymerization of olefins and transesterification. It is used as a a stabilizer PVC.They are used as solvents, lubricant additives, paint drier, and pesticides. They are used in glass coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14363-15-6
Molecular Formula C10H16O5Zn
Molecular Weight (g/mol) 281.61
MDL Number MFCD00000035
InChI Key BNHYOPPPENBGNL-SUKNRPLKSA-L
Synonym zinc 2,4-pentanedionate monohydrate
PubChem CID 122130627
IUPAC Name zinc(2+) (2Z)-pent-2-ene-2,4-bis(olate) (2Z)-4-oxopent-2-en-2-olate hydrate
SMILES O.[Zn++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=[O-]

Specifications

Melting Point 136°C to 138°C (decomposition)
Density 1.6 g/mL
Quantity 50 g
Solubility Information Soluble in alcohol; Slightly soluble in water
Formula Weight 281.60 (263.59 Anhydrous)
Physical Form Crystalline
Chemical Name or Material Zinc 2,4-pentanedionate monohydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 237-860-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.