Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc 2-ethylhexanoate, Zn ≈ 20%, cont. 1% diethylene glycolmonomethyl ether

Catalog No. aa3956022
Click to view available options
Quantity:
100 g
500 g

CAS: 136-53-8 | C16H30O4Zn | 351.792 g/mol

Zinc 2-ethylhexanoate is widely used in polyvinylchloride heat stabilizer, polyurethane catalyst and lubricant additive. It is an auxiliary drier used as pigments dispersing agent and prevents the paint film from webbing and frosting effects. It is involved in the preparation of highly porous dentritic zinc oxide nanostructures.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 136-53-8
Molecular Formula C16H30O4Zn
Molecular Weight (g/mol) 351.792
MDL Number MFCD00053316
InChI Key IFNXAMCERSVZCV-UHFFFAOYSA-L
Synonym zinc 2-ethylhexanoate, zinc bis 2-ethylhexanoate, zinc 2-ethylhexoate, hexanoic acid, 2-ethyl-, zinc salt, hexanoic acid, 2-ethyl-, zinc salt 2:1, zinc ethylhexanoate, hexanoic acid, 2-ethyl-, zinc salt, basic, zinc 2-ethylhexanoate 100g, zinc 2+ ion bis 2-ethylhexanoate
PubChem CID 61083
IUPAC Name zinc;2-ethylhexanoate
SMILES CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[Zn+2]

Specifications

Quantity 100 g
Solubility Information Immiscible with cold water.
Formula Weight 351.79
Physical Form Liquid
Chemical Name or Material Zinc 2-ethylhexanoate
Hazard Category H319-H361d
Hazard Statement GHS H Statement
H361
Suspected of damaging fertility or the unborn child.
Precautionary Statement P201-P202-P264b-P280i-P281-P305+P351+P338-P308+P313-P501c
EINECSNumber 205-251-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.