Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc, 30 Mesh, Granular, ACS, 99.8%, Spectrum™ Chemical
SDP

Catalog No. 18614714 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
Packaging:
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-614-714 12 kg Poly Pail
18-614-715 2.5 kg Poly Bottle
18-614-713 125 g Poly Bottle
18-614-716 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 18-614-714 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. Z101812KG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Chemical Identifiers

CAS 7440-66-6
Molecular Weight (g/mol) 65.39

Specifications

CAS Max % 1
Quantity 12 kg
Packaging Poly Pail
Form Granular
Percent Purity 99.8%
Grade ACS
Chemical Name or Material Zinc

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.