Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper(II) carbonate basic, pure, 54-56% Cu

Catalog No. AC196670025
Click to view available options
Quantity:
25 g
500 g
2.5 kg
Packaging:
Glass bottle

CAS: 12069-69-1 | CCuO3·CuO2H2 | 221.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12069-69-1
Molecular Formula CCuO3·CuO2H2
Molecular Weight (g/mol) 221.1
MDL Number MFCD00010976
Synonym Copper(II)hydroxide carbonate

Specifications

Melting Point 200.0°C
Color Green
Physical Form Fine Crystalline Powder
Quantity 2.5 kg
Linear Formula CuCO3·Cu(OH)2
Fieser 01,163
Merck Index 15, 2621
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in acids,ammonia,practically insoluble in alcohol
Formula Weight 221.1
Percent Purity 54 to 56% Cu
Grade Pure
Packaging Glass bottle
Density 4
Chemical Name or Material Copper(II) carbonate basic
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye irritation.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wear eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position c
EINECSNumber 235-113-6
RTECSNumber GL6910000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.