Learn More
Zinc acetate, anhydrous, 99.98% (metals basis)
Description
Zinc acetate anhydrous is used in the synthesis of layered Zn-arylphosphonates with potential application in sorption, ion exchange or catalysis. It is utilized in the ultrasonic preparation of zinc sulfide nanoparticles coated on silica particles. It is administered orally or parenterally as a nutritional supplement. It finds an application in the field of industries such as wood preservation, manufacturing other zinc salts, polymers, manufacture of ethylene acetate, as a dye mordant, and analytical reagent. It also acts as a plating inhibitor on primary water piping.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 200°C (decomposition) |
| Physical Form | Granules |
| Quantity | 10 g |
| Assay Percent Range | 99.98% (metals basis) |
| Solubility Information | Soluble in water,alcohol,dilute mineral acids and alkalies. |
| Formula Weight | 183.47 |
| Sensitivity | Hygroscopic |
| Density | 1.84 g/mL |
| Chemical Name or Material | Zinc acetate |
Safety and Handling
| Hazard Category | H302-H318 |
| Precautionary Statement | P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P501c |
| DOTInformation | Hazard Class: 9; Packaging Group: III |
| EINECSNumber | 209-170-2 |
| RTECSNumber | AK1500000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.