Learn More
Zinc acrylate
Description
The primary use of Zinc Acrylate is in the production of golf balls. Other uses are primarily in the production of specialty rubber products such as belts and hoses for automobiles. Also used as a tissue adhesive used for adherence of tissue to tissue or tissue to non-tissue for prostheses in vitro.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | ∼250°C |
| Density | 1.6 g/mL |
| Quantity | 50 g |
| UN Number | UN3077 |
| Solubility Information | Soluble completely in acrylic acid and water (as a salt). Insoluble as a polymer. |
| Formula Weight | 207.5 |
| Physical Form | Powder |
| Chemical Name or Material | Zinc acrylate |
Safety and Handling
| Hazard Category | H302-H314-H317-H335 |
| Hazard Statement | GHS H Statement H302-H315-H319-H335 Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P260-P264b-P270-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c |
| DOTInformation | Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S. |
| EINECSNumber | 238-692-3 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.