Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc-alpha-2-glycoprotein Mouse anti-Human, Clone: 4B8A4, Proteintech
SDP

Catalog No. 501736612 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6612 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6612 Supplier Proteintech Group Inc Supplier No. 661781IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

AZGP1 stimulates lipid degradation in adipocytes and causes the extensive fat losses associated with some advanced cancers. It may bind polyunsaturated fatty acids.

Specifications

Antigen Zinc-alpha-2-glycoprotein
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 4B8A4
Concentration 0.98 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene AZGP1
Gene Accession No. P25311
Gene Alias AZGP1, ZA2G, ZAG, Zinc alpha 2 glycoprotein, Zinc-alpha-2-glycoprotein, Zn alpha 2 glycoprotein, Zn alpha 2 GP, ZNGP1
Gene Symbols AZGP1
Host Species Mouse
Immunogen Zinc-alpha-2-glycoprotein Fusion Protein Ag5661
Purification Method Protein G
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 563
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.