Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc bis(trifluoromethylsulfonyl)imide

Catalog No. p-7080232
Click to view available options
Quantity:
250 mg
1 g
5 g

CAS: 168106-25-0 | C4F12N2O8S4Zn | 625.651 g/mol

Zinc bis(trifluoromethylsulfonyl)imide is used as a Catalyst for Enantioselective Friedel-Crafts alkylation of pyrroles with β-CF3 acrylates, Stereoselective Diels-Alder reaction, Tandem cyclopropane ring-opening / Conia-ene cyclization Aromatic nitration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 168106-25-0
Molecular Formula C4F12N2O8S4Zn
Molecular Weight (g/mol) 625.651
MDL Number MFCD16621474
InChI Key QEORIOGPVTWFMH-UHFFFAOYSA-N
Synonym Zinc triflimide; Zinc trifluoromethanesulfonimide
PubChem CID 131849436
IUPAC Name zinc;bis(trifluoromethylsulfonyl)azanide
SMILES C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.[Zn+2]

Specifications

Melting Point 143°C to 147°C
Color White
Flash Point >110°C (230°F)
Linear Formula [(CF3SO2)2N]2Zn
Quantity 250 mg
Sensitivity Moisture sensitive
Solubility Information It is soluble in CH2Cl2 and H2O.
Formula Weight 625.65
Physical Form Solid
Chemical Name or Material Zinc bis (trifluoromethylsulfonyl)imide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.