Alfa Aesar™ Zinc bromide, anhydrous, 98%

Manufacturer: Alfa Aesar™ B2251022

Quantity 100g

 View more versions of this product

Catalog No. AAB2251022

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7699-45-8
    Molecular Formula ZnBr2
    Formula Weight 225.19
    MDL Number MFCD00011294
    UN Number UN3260
    Merck Index 14,10129
    Chemical Name or Material Zinc bromide, anhydrous

    Specifications

    Formula Weight 225.19
    TSCA TSCA
    MDL Number MFCD00011294
    Merck Index 14,10129
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula ZnBr2
    CAS 7699-45-8
    UN Number UN3260
    Boiling Point 650°C
    Melting Point 394°C
    Quantity 100g
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
    RTECS Number ZH1150000
    Chemical Name or Material Zinc bromide, anhydrous
    Assay 98%

    Description

    Applications
    Zinc bromide solution can be used as a transparent shield against radiation. It is used as an optimal catalyst for stereospecific and regioselective reaction of silacyclopropanes with carbonyl compounds. It finds an application as Lewis acid in organic reactions.

    Solubility
    Soluble in alcohol, ether, acetone and tetrahydrofuran.

    Notes
    Hygroscopic.

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit