Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Zinc bromide, Puriss., anhydrous, ≥98%, Honeywell Fluka™
Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
500 g
1 kg
5 kg
4 x 5 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Specifications
Specifications
Physical Form | Anhydrous |
Quantity | 5 kg |
Linear Formula | ZnBr2 |
UN Number | 3260 |
Formula Weight | 225.2g/mol |
Percent Purity | ≥98% |
Grade | puriss. |
Chemical Name or Material | Zinc Bromide |
Safety and Handling
Hazard Statement | P273-P280-P305 + P351 + P338-P310 |
Precautionary Statement | H302-H314-H317-H411 |
EINECSNumber | 231-718-4 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction