Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc chloride, 0.7M solution in THF, AcroSeal™

Catalog No. p-3586518
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle

CAS: 7646-85-7 | Cl2Zn | 136.3 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 66.0°C
Chemical Name or Material Zinc chloride
Color Red
CAS 7646-85-7, 109-99-9
Physical Form Liquid
Quantity 100 mL
Molecular Formula Cl2Zn
Linear Formula ZnCl2
MDL Number MFCD00011295
Flash Point −22°C
Merck Index 15, 10331
Synonym zinc chloride, zinc dichloride, zinc chloride zncl2, zinc butter, zinc chloride fume, zinc ii chloride, zinkchloride, zintrace, zinc chloride, anhydrous, zine dichloride
Solubility Information Solubility in water: miscible
InChI Key JIAARYAFYJHUJI-UHFFFAOYSA-L
SMILES Cl[Zn]Cl
IUPAC Name dichlorozinc
Molecular Weight (g/mol) 136.3
PubChem CID 5727
ChEBI CHEBI:49976
Formula Weight 136.3
Concentration or Composition (by Analyte or Components) 0.65 to 0.8M (argentometry)
Packaging AcroSeal™ Glass Bottle
Specific Gravity 0.98
Density 0.9800g/mL
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.