Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lanthanum chloride, hydrated, 64.5-70% LaCl3
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | White |
| Physical Form | Adhering Crystals |
| Quantity | 2.5 kg |
| Linear Formula | LaCl3·xH2O |
| Merck Index | 10,5193 |
| Solubility Information | Solubility in water: soluble |
| Formula Weight | 245.27 |
| Percent Purity | 64.5 to 70% LaCl3 |
| Packaging | Glass bottle |
| Chemical Name or Material | Lanthanum chloride, hydrated |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Causes serious eye damage. May cause an allergic skin reaction. Toxic to aquatic life with long lasting effects. May be corrosive to metals. |
| Precautionary Statement | GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction