Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc Chloride, ACS Reagent, Reagents

Catalog No. 01337626
Change view
Click to view available options
Quantity:
50 g
500 g
2.5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-626 2.5 kg
01-337-625 500 g
01-337-624 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-337-626 Supplier Reagents Holdings Llc Supplier No. C13520002.5A5
Only null left
Add to Cart
Add to Cart

Zinc Chloride, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7646-85-7
Molecular Formula Cl2Zn
Molecular Weight (g/mol) 136.28
InChI Key JIAARYAFYJHUJI-UHFFFAOYSA-L
Synonym Zinc Dichloride
IUPAC Name zinc(2+) dichloride
SMILES [Cl-].[Cl-].[Zn++]

Specifications

Boiling Point 732°C
Melting Point 283°C
Color Colorless/White
pH 1
Physical Form Solid
Quantity 2.5 kg
Grade ACS
Packaging Poly Bottle
Specific Gravity 2.907
Chemical Name or Material Zinc Chloride, ACS Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.