Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Zinc Chloride, Endotoxin Tested, Granular, Multi-Compendial, U.S.P., J.T. Baker™
Shop All Avantor Products
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
Specifications
Specifications
| Physical Form | Granules |
| Quantity | 2.5 kg |
| Formula Weight | 136.3 |
| Grade | Multi-Compendial/USP |
| Packaging | Poly Bottle |
| Specific Gravity | 1L = 2.91kg |
| Chemical Name or Material | Zinc Chloride, Endotoxin Tested |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction