Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc Chloride, Saturated at 20°C, Reagents

Catalog No. 01333358
Change view
Click to view available options
:
Quantity: 4 L
Catalog No.
01-333-358 Quantity: 4 L
1 options

Catalog No. 01-333-358

Supplier: Reagents Holdings Llc CZ0019004A

Only null left
Add to Cart
Add to Cart

Zinc Chloride, Saturated at 20°C, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7646-85-7 , 7732-18-5
Molecular Formula Cl2Zn
Molecular Weight (g/mol) 136.28
InChI Key JIAARYAFYJHUJI-UHFFFAOYSA-L
Synonym Zinc Dichloride
IUPAC Name zinc(2+) dichloride
SMILES [Cl-].[Cl-].[Zn++]

Specifications

Boiling Point 120°C
Color Colorless
Physical Form Liquid
Quantity 4 L
Grade Reagent
Packaging Poly Bottle
Specific Gravity 2.1458
Chemical Name or Material Zinc Chloride, Saturated at20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.