Zinc chloride, ultra dry, 99.99% (metals basis), Alfa Aesar™

Manufacturer: Alfa Aesar 1369418

Quantity 50g

 View more versions of this product

Catalog No. AA1369418

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7646-85-7
    Physical Form Powder/Crystalline/Beads
    Assay Percent Range 99.99% metal basis
    Molecular Formula Cl2Zn
    Formula Weight 136.29
    MDL Number MFCD00011295
    Chemical Name or Material ultra dry

    Specifications

    Physical Form Powder/Crystalline/Beads
    Formula Weight 136.29
    Quantity 50g
    Color White
    Odor Odorless
    Assay Percent Range 99.99% metal basis
    Molecular Formula Cl2Zn
    MDL Number MFCD00011295
    CAS 7646-85-7
    Vapor Pressure 1hPa (1mm Hg) at 20°C
    Chemical Name or Material ultra dry

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit