Learn More
Zinc chloride, ultra dry, 99.999% (metals basis)
Description
Zinc(II) chloride is used as a catalyst, as a dehydrating and condensing agent, soldering flux and metal etchant. It is also employed in preserving anatomical specimens, wood preservatives, deodorant, disinfecting and embalming materials. It is employed as a mordant in printing and dyeing materials, and in the vulcanizing processes of fiber and rubber. It is useful as an electrolyte in dry cell batteries, in metal industry, in galvanizing iron and as an electrolyte for electroplating. It is useful in an organic synthesis, for example in the Friedel-crafts acylation, Fisher indole synthesis, Lucas reagent (zinc chloride with HCl), activation of allylic/benzylic halides towards reaction with olefins or with sodium cyanoborohydride catalyzed reduction of halides, organozinc reagents for use in Negishi coupling, and aldol condensation reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 732°C |
| Melting Point | 290°C |
| Physical Form | Powder |
| Quantity | 25 g |
| Assay Percent Range | 99.999% (metals basis) |
| Solubility Information | Freely soluble in water and dilute hydrochloric acid. Soluble in alcohol,glycerol,ether and acetone. |
| Formula Weight | 136.29 |
| Sensitivity | Hygroscopic |
| Density | 2.91 g/mL |
| Chemical Name or Material | Zinc chloride |
Safety and Handling
| Hazard Category | H302-H314-H335 |
| Precautionary Statement | P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c |
| DOTInformation | Hazard Class: 8; Packaging Group: III |
| EINECSNumber | 231-592-0 |
| RTECSNumber | ZH1400000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.