Zinc citrate dihydrate, Reagent Grade, Zn 31% min, Alfa Aesar™

Manufacturer: Alfa Aesar 1812930

Quantity 250g

 View more versions of this product

Catalog No. AA1812930

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 5990-32-9
    Physical Form Powder
    Molecular Formula C12H10O14Zn3·2H2O
    Formula Weight 610.35 (574.32 Anhydrous)
    MDL Number MFCD00150587
    Synonym Citric acid zinc salt dihydrate
    Chemical Name or Material Zinc citrate dihydrate
    Grade Reagent Grade

    Specifications

    Formula Weight 610.35 (574.32 Anhydrous)
    Quantity 250g
    EINECS Number 208-901-2
    Physical Form Powder
    MDL Number MFCD00150587
    CAS 5990-32-9
    Synonym Citric acid zinc salt dihydrate
    Molecular Formula C12H10O14Zn3·2H2O
    Chemical Name or Material Zinc citrate dihydrate
    Grade Reagent Grade

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit