Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc Copper composite, -100 mesh, 99% (metals basis)

Catalog No. AA4401322
Click to view available options
Quantity:
10 g
100 g
500 g

Catalog No. AA4401322

Supplier: Thermo Scientific Chemicals 04401322

Only null left
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Zinc Copper composite
Name Note Alloy Precursor
Avg. Mol. Wt. or Mol. Wt. Range Typically Zn 90 to 95, Cu 5 to 10 wt%
Quantity 100 g
Mesh Size −100 Mesh
Assay Percent Notes (metals basis)
MDL Number MFCD03102068
UN Number UN3089
Percent Purity 99%

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.